Novus Biologicals
Manufacturer Code:NBP273241V
Catalog # NBP273241V
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3H9 |
Immunogen | Full length recombinant protein of human PANK2 (NP_705902) produced in HEK293T cells. |
Conjugate | DyLight 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: C20orf48 EC 2.7.1.33 FLJ17232 Hallervorden-Spatz syndrome HARPFLJ11729 hPanK2 HSS MGC15053 NBIA1 pantothenate kinase 2 pantothenate kinase 2 mitochondrial Pantothenic acid kinase 2 PKANneurodegeneration with brain iron accumulation 1 (Hallervorden-Spatz syndrome); Hallervorden-Spatz syndrome; hPanK2; Pantothenate kinase 2, mitochondrial; Pantothenic acid kinase 2
Gene Aliases: C20orf48; HARP; HSS; NBIA1; PANK2; PKAN
UniProt ID: (Human) Q9BZ23
Entrez Gene ID: (Human) 80025
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.