OriGene
Manufacturer Code:CF501321
Catalog # 50-166-0344
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3H9 |
Immunogen | Full length human recombit protein of human PANK2 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: C20orf48 HARP HSS NBIA1 PKAN; Hallervorden-Spatz syndrome; hPanK2; pantothenate kinase 2 (Hallervorden-Spatz syndrome); Pantothenate kinase 2, mitochondrial; Pantothenic acid kinase 2
Gene Aliases: C20orf48; HARP; HSS; NBIA1; PANK2; PKAN
UniProt ID: (Human) Q9BZ23
Entrez Gene ID: (Human) 80025, (Dog) 477168, (Rat) 296167
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.