Invitrogen
Manufacturer Code:OSR00040W
Catalog # PIOSR00040W
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide from mouse olfactory receptor family 118 subfamily 1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Olfactory receptor 118-1; Olfactory receptor 49; Olfactory receptor I-C6; olfr49; ORL451 C6 Q8VFB7&lowbarMOUSE Olfactory receptor MOR118-1 Olfactory receptor Olfr49. GENE NAME: Olfr49
Gene Aliases: IC6; Mor118-1; MOR1181; Olfr49
UniProt ID: (Mouse) Q9Z1V0
Entrez Gene ID: (Mouse) 18348
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.