Novus Biologicals
Manufacturer Code:NBP219648
Catalog # NBP219648
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human OXSM. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-ketoacyl-acyl carrier protein synthase; 3-ketoacyl-acyl carrier protein synthase 3-oxoacyl-[acyl-carrier-protein] synthase mitochondrial 3-oxoacyl-ACP synthase mitochondrial acyl-malonyl acyl carrier protein-condensing enzyme beta-ketoacyl synthase Beta-ketoacyl-ACP synthase EC 2.3.1 EC 2.3.1.41 FASN2D FLJ20604 KASI KS3-oxoacyl-acyl-carrier-protein synthase type II mitochondrial beta-ketoacyl synthase; 3-oxoacyl-[acyl-carrier-protein] synthase, mitochondrial; 3-oxoacyl-acyl-carrier-protein synthase; acyl-malonyl acyl carrier protein-condensing enzyme; beta-ketoacyl synthase; Beta-ketoacyl-ACP synthase; type II mitochondrial beta-ketoacyl synthase
Gene Aliases: FASN2D; KASI; KS; OXSM
UniProt ID: (Human) Q9NWU1
Entrez Gene ID: (Human) 54995
Molecular Function: acyltransferase dehydrogenase esterase hydrolase ligase methyltransferase oxidoreductase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.