Invitrogen
Manufacturer Code:PA595086
Catalog # PIPA595086
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence in the middle region of human OSBP1 identical to the related rat and mouse sequences. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Oxysterol-binding protein 1; Oxysterol-binding protein 1 OSBP OSBP1
Gene Aliases: 1110018F06Rik; AW559088; Kiaa4220; mKIAA4220; OSBP; OSBP1
UniProt ID: (Human) P22059, (Mouse) Q3B7Z2
Entrez Gene ID: (Human) 5007, (Mouse) 76303, (Rat) 365410
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.