Abnova
Manufacturer Code:H00004987K
Catalog # 89-205-779
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human OPRL1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human OPRL1 peptide using ARM Technology.
For Research Use Only
Protein Aliases: Kappa-type 3 opioid receptor; kappa3-related opioid receptor; KOR-3; KOR-3 MGC34578 NOCIR OOR ORL1; Nociceptin receptor; nociceptin/orphanin FQ receptor; opiate receptor-like 1; Orphanin FQ receptor
Gene Aliases: KOR-3; NOCIR; NOP; NOPr; OOR; OPRL1; ORL1
UniProt ID: (Human) P41146
Entrez Gene ID: (Human) 4987
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.