Novus Biologicals
Manufacturer Code:NB11058849
Catalog # NB11058849
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoblot (IB) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to an internal region of the human NOX4 protein (between residues 100-200) [UniProt Q9NPH5]. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 1.6.3 Kidney oxidase-1 Kidney superoxide-producing NADPH oxidase KOX KOX-1Renal NAD(P)H-oxidase NADPH oxidase 4 RENOXEC 1.6.3.-; Kidney oxidase-1; Kidney superoxide-producing NADPH oxidase; KOX-1; NADPH oxidase 4; Renal NAD(P)H-oxidase
Gene Aliases: KOX; KOX-1; NOX4; RENOX
UniProt ID: (Human) Q9NPH5
Entrez Gene ID: (Human) 50507
Molecular Function: oxidase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.