Novus Biologicals
Manufacturer Code:NBP226214
Catalog # NBP226214
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-TYHTVLQTSADFID corresponding to internal region according to NP_000262.2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ98532 Niemann-Pick C1 protein Niemann-Pick disease type C1 NPC NPC1; Niemann-Pick C1 protein; Niemann-Pick disease, type C1; NPC intracellular cholesterol transporter 1; truncated Niemann-Pick C1
Gene Aliases: NPC; NPC1
UniProt ID: (Human) O15118
Entrez Gene ID: (Human) 4864
Molecular Function: receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.