Invitrogen
Manufacturer Code:PA576740
Catalog # PIPA576740
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human NQO2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DHQVDIA6EC 1.10.99.2MGC94180NAD(P)H dehydrogenase quinone 2NAD(P)H menadione oxidoreductase 1 dioxin inducible 2NAD(P)H menadione oxidoreductase 2 dioxin inducibleNMOR2NQO 2NQO2NQO2&lowbarHUMANNRH dehydrogenase &lsqbquinone] 2NRH dehydrogenaseNRH:quinone oxidoreductase 2OTTHUMP00000015948OTTHUMP00000015949OTTHUMP00000015953Ox 2Ox2QR2QuinoneQuinone reductase 2Ribosyldihydronicotinamide dehydrogenase &lsqbquinone]Ribosyldihydronicotinamide dehydrogenase; NAD(P)H dehydrogenase, quinone 2; NAD(P)H menadione oxidoreductase 2, dioxin inducible; NAD(P)H menadione oxidoreductase-1, dioxin-inducible-2; NRH dehydrogenase [quinone] 2; NRH: quinone oxidoreductase; NRH:quinone oxidoreductase 2; QR2; Quinone reductase 2; ribosyldihydronicotinamide dehydrogenase; Ribosyldihydronicotinamide dehydrogenase [quinone]
Gene Aliases: DHQV; DIA6; NMO2; NMOR2; NQO2; Ox2; QR2
UniProt ID: (Human) P16083, (Mouse) Q9JI75
Entrez Gene ID: (Human) 4835, (Mouse) 18105
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.