Invitrogen
Manufacturer Code:PA533991
Catalog # PIPA533991
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 16 amino acid peptide from 3rd cytoplasmic domain of human NPY5R. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Neuropeptide Y receptor type 5; NPY-Y5 receptor; NPY5-R; NPY5R NPYR5 NPY5-R NPYY5 Neuropeptide Y receptor Y5 NPYY5-R Neuropeptide Y receptor type 5 Neuropeptide Y5 receptor NPY-Y5 receptor NPY5 receptor Y5 receptor; NPYY5-R; Y5 receptor
Gene Aliases: NPY5-R; NPY5R; NPYR5; NPYY5-R
UniProt ID: (Human) Q15761
Entrez Gene ID: (Human) 4889
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.