R&D Systems
Manufacturer Code:MAB6400SP
Catalog # MAB6400SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 556153 |
Immunogen | NS0 mouse myeloma cell line transfected with human NPY1R Met1-Ile384 Accession # P25929 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Neuropeptide Y receptor type 1; neuropeptide Y receptor type 1 neuropeptide Y receptor Y1 NPY1-R NPYR NPYY1; NPY1-R
Gene Aliases: NPY1-R; NPY1R; NPYR; NPYY1
UniProt ID: (Human) P25929
Entrez Gene ID: (Human) 4886
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.