Novus Biologicals
Manufacturer Code:NBP299830F
Catalog # NB089716
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant NLK (Q9UBE8 Val121-Glu527). |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp761G1211 EC 2.7.11 EC 2.7.11.24 FLJ21033 LAK1 nemo like kinase nemo-like kinaseserine/threonine-protein kinase NLK Protein LAK1; Nemo-like kinase; Protein LAK1; Serine/threonine-protein kinase NLK
Gene Aliases: LAK1; NLK
UniProt ID: (Human) Q9UBE8
Entrez Gene ID: (Human) 51701
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.