R&D Systems
Manufacturer Code:FAB8404G100UG
Catalog # FAB8404G100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 896151 |
Immunogen | NS0 mouse myeloma cell line transfected with human NHA2/SLC9B2 Met1-Val537 Accession # NP_849155 |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: FLJ23984 Mitochondrial Na(+)/H(+) exchanger NHA2 mitochondrial sodium/hydrogen exchanger NHA2 Na(+)/H(+) exchanger-like domain-containing protein 2 Na+/H+ exchanger domain containing 2 NHA2NHE10 NHE domain-containing protein 2 NHE10 NHEDC2 SLC9B2 Sodium/hydrogen exchanger-like domain-containing protein 2; mitochondrial Na(+)/H(+) exchanger NHA2; mitochondrial sodium/hydrogen exchanger NHA2; Na(+)/H(+) exchanger NHA2; Na(+)/H(+) exchanger-like domain-containing protein 2; Na+/H+ exchanger domain containing 2; NHE domain-containing protein 2; Sodium/hydrogen exchanger 9B2; Sodium/hydrogen exchanger-like domain-containing protein 2; Solute carrier family 9 subfamily B member 2; solute carrier family 9, subfamily B (cation proton antiporter 2), member 2; solute carrier family 9, subfamily B (NHA2, cation proton antiporter 2), member 2
Gene Aliases: NHA2; NHE10; NHEDC2; SLC9B2
UniProt ID: (Human) Q86UD5
Entrez Gene ID: (Human) 133308
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.