Invitrogen
Manufacturer Code:PA587249
Catalog # PIPA587249
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human NF2/Merlin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Merlin; Merlin Moesin-ezrin-radixin-like protein Neurofibromin-2 Schwannomerlin Schwannomin NF2 SCH; moesin-ezrin-radixin like; Moesin-ezrin-radixin-like protein; moesin-ezrin-radizin-like protein; neurofibromatosis 2; neurofibromin 2 (bilateral acoustic neuroma); neurofibromin 2 (merlin); Neurofibromin-2; Schwannomerlin; Schwannomin
Gene Aliases: ACN; BANF; merlin; Nf-2; NF2; SCH
UniProt ID: (Human) P35240, (Mouse) P46662, (Rat) Q63648
Entrez Gene ID: (Human) 4771, (Mouse) 18016, (Rat) 25744
Molecular Function: actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.