Novus Biologicals
Manufacturer Code:NBP128863
Catalog # NBP128863
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 7A7A9 |
Immunogen | Purified recombinant fragment of NCOR1(aa1-192) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: hN-CoR KIAA1047N-CoR1 MGC104216 N-CoRhCIT529I10 nuclear receptor corepressor 1 nuclear receptor co-repressor 1 thyroid hormone- and retinoic acid receptor-associated corepressor 1 TRAC1; N-CoR; Nuclear receptor corepressor 1; protein phosphatase 1, regulatory subunit 109; thyroid hormone- and retinoic acid receptor-associated corepressor 1
Gene Aliases: hN-CoR; KIAA1047; N-CoR; N-CoR1; NCOR1; PPP1R109; TRAC1
UniProt ID: (Human) O75376
Entrez Gene ID: (Human) 9611
Molecular Function: nucleic acid binding transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.