R&D Systems
Manufacturer Code:AF6167SP
Catalog # AF6167SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human NCOR1 Gln1770-Ala1947 Accession # O75376 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: hN-CoR KIAA1047 KIAA1047N-CoR1 MGC104216 N-CoR N-CoRhCIT529I10 nuclear receptor corepressor 1 nuclear receptor co-repressor 1 thyroid hormone- and retinoic acid receptor-associated corepressor 1 TRAC1; N-CoR; Nuclear receptor corepressor 1; protein phosphatase 1, regulatory subunit 109; thyroid hormone- and retinoic acid receptor-associated corepressor 1
Gene Aliases: hN-CoR; KIAA1047; N-CoR; N-CoR1; NCOR1; PPP1R109; TRAC1
UniProt ID: (Human) O75376
Entrez Gene ID: (Human) 9611
Molecular Function: nucleic acid binding transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.