Novus Biologicals
Manufacturer Code:NBP297690IR
Catalog # NB046603
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human MSS4 fragment. |
Conjugate | DyLight 755 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.1 MSS4Protein STM-7 Phosphatidylinositol-4-phosphate 5-kinase type I beta phosphatidylinositol-4-phosphate 5-kinase type-1 beta phosphatidylinositol-4-phosphate 5-kinase type I beta PIP5K1-beta PIP5KIbeta PtdIns(4)P-5-kinase 1 beta STM7EC 2.7.1.68 Type I phosphatidylinositol-4-phosphate 5-kinase beta; Phosphatidylinositol 4-phosphate 5-kinase type I beta; Phosphatidylinositol 4-phosphate 5-kinase type-1 beta; phosphatidylinositol-4-phosphate 5-kinase type-1 beta; phosphatidylinositol-4-phosphate 5-kinase, type I, beta; PIP5K1-beta; PIP5KIbeta; Protein STM-7; ptdIns(4)P-5-kinase 1 beta; Type I phosphatidylinositol 4-phosphate 5-kinase beta; type I phosphatidylinositol-4-phosphate 5-kinase beta
Gene Aliases: MSS4; PIP5K1B; STM7
UniProt ID: (Human) B4DIG7
Entrez Gene ID: (Human) 8395
Molecular Function: kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.