OriGene
Manufacturer Code:CF505299
Catalog # 50-166-3531
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1E5 |
Immunogen | Full length human recombit protein of human MRPL11 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 39S ribosomal protein L11, mitochondrial; CGI-113 L11MT MRP-L11; L11mt; Mitochondrial large ribosomal subunit protein uL11m
Gene Aliases: CGI-113; L11MT; MRP-L11; MRPL11
UniProt ID: (Human) Q9Y3B7
Entrez Gene ID: (Human) 65003
Molecular Function: RNA binding protein nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.