Novus Biologicals
Manufacturer Code:NBP237347
Catalog # NBP237347
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 6A7H3 |
Immunogen | Purified recombinant fragment of human ABCC4 (AA: 631-692) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ATP-binding cassette sub-family C member 4; ATP-binding cassette sub-family C member 4 ATP-binding cassette sub-family C (CFTR/MRP) member 4 EST170205 MOAT-BbA464I2.1 (ATP-binding cassette sub-family C (CFTR/MRP) member 4) MOATBcanalicular multispecific organic anion transporter (ABC superfamily) MRP4MRP/cMOAT-related ABC transporter multidrug resistance-associated protein 4 multispecific organic anion transporter B Multi-specific organic anion transporter B; bA464I2.1 (ATP-binding cassette, sub-family C (CFTR/MRP), member 4); canalicular multispecific organic anion transporter (ABC superfamily); MOAT-B; MRP/cMOAT-related ABC transporter; Multi-specific organic anion transporter B; Multidrug resistance-associated protein 4
Gene Aliases: ABCC4; MOAT-B; MOATB; MRP4
UniProt ID: (Human) O15439
Entrez Gene ID: (Human) 10257
Molecular Function: ATP-binding cassette (ABC) transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.