Invitrogen
Manufacturer Code:PA5106625
Catalog # PIPA5106625
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ABCC2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABC30 ABCC2 AI173996 ATP binding cassette subfamily C member 2 ATP-binding cassette sub-family C member 2 ATP-binding cassette subfamily C (CFTR/MRP) member 2 ATP-binding cassette sub-family C (CFTR/MRP) member 2 calicular multispecific organic anion transporter Canalicular multidrug resistance protein Canalicular multispecific organic anion transporter 1 CMOAT CMOAT1 CMRP DJS Human MRP2 KIAA1010 MRP2 multidrug resistance associated protein 2 multidrug resistance protein 2 multidrug resistance-associated protein 2; ATP-binding cassette sub-family C member 2; ATP-binding cassette, sub-family C (CFTR/MRP), member 2; calicular multispecific organic anion transporter; Canalicular multidrug resistance protein; Canalicular multispecific organic anion transporter 1; multidrug resistance associated protein 2; multidrug resistance protein 2; Multidrug resistance-associated protein 2
Gene Aliases: ABC30; ABCC2; AI173996; CMOAT; CMOAT1; CMRP; DJS; MRP2
UniProt ID: (Human) Q92887, (Rat) Q63120, (Mouse) Q8VI47
Entrez Gene ID: (Human) 1244, (Rat) 25303, (Mouse) 12780
Molecular Function: ATP-binding cassette (ABC) transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.