Proteintech
Manufacturer Code:115451AP150UL
Catalog # 50-172-7253
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MOXD1 Fusion Protein Ag2060 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DBH-like monooxygenase protein 1; DBH-related protein; dopamine-oxygenase; Monooxygenase X; Monooxygenase X monooxygenase DBH like 1 MOX MOXD1 PRO5780; monooxygenase, DBH-like 1
Gene Aliases: 3230402N08Rik; Dbhr; dJ248E1.1; MNCb-5203; MOX; MOXD1; PRO5780; UNQ2493/PRO5780
UniProt ID: (Human) Q6UVY6, (Mouse) Q9CXI3
Entrez Gene ID: (Human) 26002, (Mouse) 59012, (Rat) 294119
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.