Invitrogen
Manufacturer Code:PA5103309
Catalog # PIPA5103309
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human MAP3K9 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E130314H24Rik Map3k9 MEKK9 mitogen activated protein kinase kinase kinase 9 mitogen-activated protein kinase kinase kinase 9 Mixed lineage kinase 1 mixed lineage kinase 1 (tyr and ser/thr specificity) mixed-lineage kinase 1 MLK1 Prke1 RGD1562149; mitogen activated protein kinase kinase kinase 9; Mitogen-activated protein kinase kinase kinase 9; Mixed lineage kinase 1; mixed lineage kinase 1 (tyr and ser/thr specificity)
Gene Aliases: E130314H24Rik; MAP3K9; MEKK9; MLK1; PRKE1
UniProt ID: (Human) P80192, (Mouse) Q3U1V8
Entrez Gene ID: (Human) 4293, (Mouse) 338372
Molecular Function: kinase non-receptor serine/threonine protein kinase non-receptor tyrosine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.