Invitrogen
Manufacturer Code:PA579628
Catalog # PIPA579628
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the N-terminus of human MEK7. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: c-Jun N-terminal kinase kinase 2; Dual specificity mitogen-activated protein kinase kinase 7; Dual specificity mitogen-activated protein kinase kinase 7 MAP kinase kinase 7 MAPKK 7 JNK-activating kinase 2 MAPK/ERK kinase 7 MEK 7 Stress-activated protein kinase kinase 4 SAPK kinase 4 SAPKK-4 SAPKK4 c-Jun N-terminal kinase kinase 2 JNK kinase 2 JNKK 2 MAP2K7 JNKK2 MEK7 MKK7 PRKMK7 SKK4; JNK kinase 2; JNK-activating kinase 2; JNKK 2; MAP kinase kinase 7; map2k7; map2k7 {ECO:0000312|EMBL:AAX61178.1}; MAPK/ERK kinase 7; MAPKK 7; MEK 7; mitogen activated protein kinase kinase 7; protein kinase, mitogen activated, kinase 7; SAPK kinase 4; Stress-activated protein kinase kinase 4
Gene Aliases: JNKK; 2; 5930412N11Rik; JNKK 2; JNKK2; MAP2K7; MAPKK 7; MAPKK7; MEK; MEK 7; MEK7; MKK7; PRKMK7; SAPKK-4; SAPKK4; sek2; SKK4
UniProt ID: (Human) O14733, (Rat) Q4KSH7, (Mouse) Q8CE90
Entrez Gene ID: (Human) 5609, (Rat) 363855, (Mouse) 26400
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.