Invitrogen
Manufacturer Code:PA560242
Catalog # PIPA560242
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human MFSD2A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Major facilitator superfamily domain-containing protein 2A; mfsd2a zgc:101615 major facilitator superfamily domain containing 2a major facilitator superfamily domain-containing protein 2A-A; NLS1; sodium-dependent LPC symporter 1; Sodium-dependent lysophosphatidylcholine symporter 1
Gene Aliases: HMFN0656; MCPH15; MFSD2; MFSD2A; NLS1; PP9177; UNQ300/PRO341
UniProt ID: (Human) Q8NA29
Entrez Gene ID: (Human) 84879
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.