Invitrogen
Manufacturer Code:PA521550
Catalog # PIPA521550
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 161 and 519 of (Uniprot IDNo.P48163) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cytosolic malic enzyme 1; HUMNDME MES Malic enzyme cytoplasmic NADP-ME NADP-dependent malic enzyme malate dehydrogenase malic enzyme 1 soluble pyruvic-malic carboxylase; malate dehydrogenase; malate dehydrogenase (oxaloacetate-decarboxylating) (NADP(+)); malate dehydrogenase (oxaloacetate-decarboxylating) (NADP+); Malic enzyme 1; malic enzyme 1, NADP(+)-dependent, cytosolic; malic enzyme 1, soluble; Malic enzyme, cytoplasmic; malic enzyme, supernatant; NADP-dependent malic enzyme; NADP-ME; pyruvic-malic carboxylase
Gene Aliases: D9Ertd267e; HUMNDME; Mdh-1; ME1; MES; Mod-1; MOD1
UniProt ID: (Human) P48163, (Mouse) P06801
Entrez Gene ID: (Human) 4199, (Mouse) 17436, (Rat) 24552
Molecular Function: acyltransferase decarboxylase dehydrogenase lyase oxidoreductase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.