Invitrogen
Manufacturer Code:MA524082
Catalog # PIMA524082
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 345404 |
Immunogen | Y3 rat myeloid cell line transfeced with mouse Melanocortin 3R/MC3R |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: MC3-R; MC3R BMIQ9 MC3 receptor Melanocortin receptor 3 OB20 MC3 MC3-R Melanocortin 3 receptor OQTL; Melanocortin receptor 3
Gene Aliases: MC3-R; Mc3r
UniProt ID: (Mouse) P33033
Entrez Gene ID: (Mouse) 17201
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.