Invitrogen
Manufacturer Code:PA572562
Catalog # PIPA572562
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 86-112 amino acids from the Central region of human MC3R |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MC3-R; MC3R BMIQ9 MC3 receptor Melanocortin receptor 3 OB20 MC3 MC3-R Melanocortin 3 receptor OQTL; Melanocortin receptor 3; obesity quantitative trait locus
Gene Aliases: BMIQ9; MC3; MC3-R; MC3R; OB20; OQTL
UniProt ID: (Human) P41968
Entrez Gene ID: (Human) 4159
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.