Invitrogen
Manufacturer Code:PA533924
Catalog # PIPA533924
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 18 amino acid peptide from C-Terminus of human MC3 Receptor. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MC3-R; MC3R BMIQ9 MC3 receptor Melanocortin receptor 3 OB20 MC3 MC3-R Melanocortin 3 receptor OQTL; Melanocortin receptor 3; obesity quantitative trait locus
Gene Aliases: BMIQ9; MC3; MC3-R; MC3R; OB20; OQTL
UniProt ID: (Human) P41968
Entrez Gene ID: (Human) 4159
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.