Invitrogen
Manufacturer Code:PA5101199
Catalog # PIPA5101199
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human MAP4K3 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4833416M01Rik 4833416M07Rik 9530052P13Rik germinal center kinase-like kinase germinal center kinase-related protein kinase GLK MAP4K3 MAPK/ERK kinase kinase kinase 3 MAPKKKK3 MEK kinase kinase 3 MEKKK 3 MEKKK3 mitogen activated protein kinase kinase kinase kinase 3 mitogen-activated protein kinase kinase kinase kinase 3 RAB8IPL1; germinal center kinase-like kinase; Germinal center kinase-related protein kinase; GLK; MAPK/ERK kinase kinase kinase 3; MEK kinase kinase 3; MEKKK 3; mitogen activated protein kinase kinase kinase kinase 3; Mitogen-activated protein kinase kinase kinase kinase 3
Gene Aliases: 4833416M01Rik; 4833416M07Rik; 9530052P13Rik; GLK; MAP4K3; MAPKKKK3; MEKKK 3; MEKKK3; RAB8IPL1
UniProt ID: (Human) Q8IVH8, (Mouse) Q99JP0, (Rat) Q924I2
Entrez Gene ID: (Human) 8491, (Mouse) 225028, (Rat) 170920
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.