Invitrogen
Manufacturer Code:PA552422
Catalog # PIPA552422
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human MAP4K2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B lymphocyte serine/threonine protein kinase; B lymphocyte serine/threonine-protein kinase; GC kinase; Germinal center kinase; germinal centre kinase (GC kinase); MAPK/ERK kinase kinase kinase 2; MEK kinase kinase 2; MEKKK 2; Mitogen-activated protein kinase kinase kinase kinase 2; Mitogen-activated protein kinase kinase kinase kinase 2 MAPK/ERK kinase kinase kinase 2 MEK kinase kinase 2 MEKKK 2 Germinal center kinase GC kinase Rab8 interacting protein B lymphocyte serine/threonine-protein kinase; Rab8 interacting protein; Rab8-interacting protein
Gene Aliases: BL44; GCK; MAP4K2; RAB8IP
UniProt ID: (Human) Q12851
Entrez Gene ID: (Human) 5871
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.