Proteintech
Manufacturer Code:552441AP150UL
Catalog # 50-173-6078
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B lymphocyte serine/threonine protein kinase; B lymphocyte serine/threonine-protein kinase; BL44 GC kinase GCK Germinal center kinase MAP4K2 MEK kinase kinase 2 MEKKK 2 Rab8 interacting protein RAB8IP; GC kinase; GCK; Germinal center kinase; germinal centre kinase (GC kinase); MAPK/ERK kinase kinase kinase 2; MEK kinase kinase 2; MEKKK 2; mitogen activated protein kinase kinase kinase kinase 2; Mitogen-activated protein kinase kinase kinase kinase 2; RAB8 interacting protein; Rab8-interacting protein
Gene Aliases: AI385662; BL44; GCK; MAP4K2; RAB8IP
UniProt ID: (Human) Q12851, (Mouse) Q61161
Entrez Gene ID: (Human) 5871, (Rat) 293694, (Mouse) 26412
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.