Proteintech
Manufacturer Code:216101AP150UL
Catalog # 50-173-3512
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MAP3K4 Fusion Protein Ag16254 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dJ473J16.1 (mitogen-activated protein kinase kinase kinase 4); KIAA0213 MAP three kinase 1 MAP3K4 MAPK/ERK kinase kinase 4 MAPKKK4 MEK kinase 4 MEKK 4 MEKK4 MTK1 PRO0412; MAP three kinase 1; MAP/ERK kinase kinase 4; MAPK/ERK kinase kinase 4; MEK kinase 4; mitogen activated protein kinase kinase kinase 4; Mitogen-activated protein kinase kinase kinase 4; predicted protein of HQ0412; SSK2/SSK22 MAP kinase kinase kinase, yeast, homolog of; T-associated sex reversal
Gene Aliases: D17Rp17; D17Rp17e; KIAA0213; MAP3K4; MAPKKK4; Mek4b; MEKK 4; MEKK4; mKIAA0213; MTK1; PRO0412; RP17; Rp17a; Tas
UniProt ID: (Human) Q9Y6R4, (Mouse) O08648
Entrez Gene ID: (Human) 4216, (Mouse) 26407, (Rat) 308106
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.