Invitrogen
Manufacturer Code:MA521649
Catalog # PIMA521649
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2A2 |
Immunogen | LHX6 (NP&lowbar055183 274 a.a. &bsim 363 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: LIM homeobox protein 6; LIM homeodomain protein 6.1; LIM/homeobox protein Lhx6; LIM/homeobox protein Lhx6.1; RP11-498E2.6 LHX6.1 MGC119542 MGC119544 MGC119545 LIM homeobox protein 6 LIM homeodomain protein 6.1 LIM/homeobox protein Lhx6 LIM/homeobox protein Lhx6.1
Gene Aliases: LHX6; LHX6.1
UniProt ID: (Human) Q9UPM5
Entrez Gene ID: (Human) 26468
Molecular Function: RNA binding protein actin family cytoskeletal protein cytoskeletal protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.