Invitrogen
Manufacturer Code:PA544919
Catalog # PIPA544919
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human LYPLA2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acyl-protein thioesterase 2; APT-2; LPL-II; Lysophospholipase 2; Lysophospholipase II; lysoPLA II; mLyso II; testicular tissue protein Li 3
Gene Aliases: APT-2; APT2; DJ886K2.4; LYPLA2; LysoII
UniProt ID: (Human) Q7Z4Z2, (Rat) Q9QYL8, (Mouse) Q9WTL7
Entrez Gene ID: (Dog) 100855579, (Bovine) 784764, (Human) 11313, (Horse) 100071559, (Rabbit) 100345346, (Rat) 83510, (Mouse) 26394, (Guinea pig) 100713567
Molecular Function: hydrolase lipase phospholipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.