Invitrogen
Manufacturer Code:PA599926
Catalog # PIPA599926
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human RXFP1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Gm1018 leucine-rich repeat-containing 7 leucine-rich repeat-containing G protein-coupled receptor 7 Leucine-rich repeat-containing G-protein coupled receptor 7 LGR7 LGR7.1 LGR7.10 LGR7.2 MGC138347 MGC142177 Relaxin family peptide receptor 1 relaxin receptor 1 relaxin/insulin like family peptide receptor 1 relaxin/insulin-like family peptide receptor 1 RXFP1 RXFPR1; leucine-rich repeat-containing 7; leucine-rich repeat-containing G protein-coupled receptor 7; Leucine-rich repeat-containing G-protein coupled receptor 7; Relaxin family peptide receptor 1; Relaxin receptor 1; relaxin/insulin-like family peptide receptor 1
Gene Aliases: Gm1018; LGR7; RXFP1; RXFPR1
UniProt ID: (Human) Q9HBX9, (Rat) Q6R6I6, (Mouse) Q6R6I7
Entrez Gene ID: (Human) 59350, (Rat) 295144, (Mouse) 381489
Molecular Function: extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.