Invitrogen
Manufacturer Code:PA597919
Catalog # PIPA597919
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human Plastin-2 protein (2-627AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 65 kDa macrophage protein; bA139H14.1 (lymphocyte cytosolic protein 1 (L-plastin)); epididymis secretory protein Li 37; L-fimbrin; L-plastin; L-plastin (Lymphocyte cytosolic protein 1) (LCP-1) (65 kDa macrophage protein) (PP65); L-plastin (Lymphocyte cytosolic protein 1) (LCP-1) (LC64P); LC64P; LCP-1; Lymphocyte cytosolic protein 1; lymphocyte cytosolic protein 1 (L-plastin); Lymphocyte cytosolic protein-1 (plasmin); plastin 2; plastin 2, L; Plastin-2; pp65
Gene Aliases: AW536232; CP64; D14Ertd310e; HEL-S-37; L-PLASTIN; LC64P; LCP-1; LCP1; LPL; PLS2; pp65
UniProt ID: (Human) P13796, (Mouse) Q61233
Entrez Gene ID: (Human) 3936, (Mouse) 18826
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.