Novus Biologicals
Manufacturer Code:NBP237538
Catalog # NBP237538
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5000000000000 |
Immunogen | Purified recombinant fragment of human KCNQ1 expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ATFB3 IKs producing slow voltage-gated potassium channel subunit alpha KvLQT1 JLNS1 KCNA8FLJ26167 KCNA9Kv1.9 kidney and cardiac voltage dependend K+ channel KQT-like 1 Kv7.1 KVLQT1ATFB1 LQT LQT1 potassium voltage-gated channel subfamily KQT member 1 potassium voltage-gated channel KQT-like subfamily member 1 RWS slow delayed rectifier channel subunit SQT2 Voltage-gated potassium channel subunit Kv7.1 WRS; IKs producing slow voltage-gated potassium channel subunit alpha KvLQT1; kidney and cardiac voltage dependend K+ channel; KQT-like 1; potassium channel, voltage gated KQT-like subfamily Q, member 1; Potassium voltage-gated channel subfamily KQT member 1; potassium voltage-gated channel, KQT-like subfamily, member 1; slow delayed rectifier channel subunit; Voltage-gated potassium channel subunit Kv7.1
Gene Aliases: ATFB1; ATFB3; JLNS1; KCNA8; KCNA9; KCNQ1; Kv1.9; Kv7.1; KVLQT1; LQT; LQT1; RWS; SQT2; WRS
UniProt ID: (Human) P51787
Entrez Gene ID: (Human) 3784
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.