Proteintech
Manufacturer Code:244091AP150UL
Catalog # 50-173-4338
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KTELC1 Fusion Protein Ag19942 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 9630046K23Rik; C3orf9 CAP10 like 46 kDa protein CLP46 hCLP46 KDELCL1 KTELC1 MDS010 MDSRP UNQ490/PRO1006; CAP10-like 46 kDa protein; hCLP46; hRumi; KDELC family like 1; KTEL (Lys-Tyr-Glu-Leu) containing 1; KTEL motif-containing protein 1; Myelodysplastic syndromes relative protein; O-glucosyltransferase Rumi homolog; Protein O-glucosyltransferase 1; protein O-xylosyltransferase; Protein O-xylosyltransferase POGLUT1; x 010 protein
Gene Aliases: C3orf9; CLP46; hCLP46; KDELCL1; KTELC1; MDS010; MDSRP; POGLUT1; Rumi; UNQ490/PRO1006
UniProt ID: (Human) Q8NBL1
Entrez Gene ID: (Human) 56983
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.