Abnova
Manufacturer Code:PAB8799
Catalog # 89-133-944
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to residues surrounding amino acids 17 of mouse Kiss1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Kiss1.
For Research Use Only
Protein Aliases: 0610012C01Rik; Kiss1 metastasis-suppressor; Kisspeptin-1; Kisspeptin-10; Kisspeptin-52; Metastasis-suppressor KiSS-1; metastatin; Metastin; Metastin45-54
Gene Aliases: Kiss1; kisspeptin; metastatin
UniProt ID: (Mouse) Q6Y4S4
Entrez Gene ID: (Mouse) 280287
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.