R&D Systems
Manufacturer Code:AF7028SP
Catalog # AF7028SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant mouse Inositol Monophosphatase 3/IMPAD1 Asn145-His356 Accession # Q80V26 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: 3'(2'), 5'-bisphosphate nucleotidase 2; EC 3.1.3 EC 3.1.3.25 FLJ20421 IMP 3 IMP3 IMPA3 IMPA3IMPase 3 IMPase 3 inositol monophosphatase 3 inositol monophosphatase domain containing 1 Inositol monophosphatase domain-containing protein 1 Inositol-1(or 4)-monophosphatase 3 Myo-inositol monophosphatase A3; Golgi 3-prime phosphoadenosine 5-prime phosphate 3-prime phosphatase; Golgi-resident adenosine 3',5'-bisphosphate 3'-phosphatase; golgi-resident nucleotide phosphatase; Golgi-resident PAP phosphatase; IMPase 3; Inositol monophosphatase domain-containing protein 1; inositol-1(or 4)-monophosphatase 3; Myo-inositol monophosphatase A3; Phosphoadenosine phosphate 3'-nucleotidase
Gene Aliases: BPNT2; GPAPP; IMP 3; IMP-3; IMPA3; IMPAD1
UniProt ID: (Human) Q9NX62
Entrez Gene ID: (Human) 54928
Molecular Function: hydrolase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.