Invitrogen
Manufacturer Code:PA579523
Catalog # PIPA579523
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E.coli-derived mouse ISG15 recombit protein (Position: A2-G155). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hUCRP; interferon, alpha-inducible protein; interferon, alpha-inducible protein (clone IFI-15K); Interferon-induced 15 kDa protein; interferon-induced 15-KDa protein; Interferon-induced 17 kDa protein; interferon-induced 17-kDa/15-kDa protein; interferon-stimulated protein (15 kDa); interferon-stimulated protein 15; interferon-stimulated protein, 15 kDa; IP17; Ubiquitin cross-reactive protein; Ubiquitin-like protein ISG15; Ubiquitin-like protein ISG15 Interferon-induced 15 kDa protein Interferon-induced 17 kDa protein IP17 Ubiquitin cross-reactive protein Isg15 G1p2 Ucrp
Gene Aliases: G1P2; hUCRP; IFI15; IGI15; IMD38; IP17; Irfp; ISG15; UCRP
UniProt ID: (Human) P05161, (Mouse) Q64339
Entrez Gene ID: (Human) 9636, (Mouse) 100038882
Molecular Function: RNA binding protein nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.