Invitrogen
Manufacturer Code:PA576968
Catalog # PIPA576968
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human INPP5J. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Inositol polyphosphate 5-phosphatase J; phosphatidylinositol (4,5) bisphosphate 5-phosphatase, A; Phosphatidylinositol 4,5-bisphosphate 5-phosphatase A; Phosphatidylinositol 45-bisphosphate 5-phosphatase A Inositol polyphosphate 5-phosphatase J INPP5J PIB5PA PIPP; Proline-rich inositol polyphosphate 5-phosphatase
Gene Aliases: Inpp; INPP5; INPP5J; PIB5PA; PIPP
UniProt ID: (Human) Q15735, (Mouse) P59644, (Rat) Q9JMC1
Entrez Gene ID: (Human) 27124, (Mouse) 170835, (Rat) 171088
Molecular Function: hydrolase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.