Proteintech
Manufacturer Code:151411AP150UL
Catalog # 50-173-0016
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | INPP5B Fusion Protein Ag7315 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5PTase; 5PTase INPP5B Phosphoinositide 5 phosphatase; 75 kDa inositol polyphosphate-5-phosphatase; Inositol polyphosphate-5-phosphatase B; inositol polyphosphate-5-phosphatase, 75kDa; Phosphoinositide 5-phosphatase; Type II inositol 1,4,5-trisphosphate 5-phosphatase
Gene Aliases: 5PTase; AW260155; INPP5B; INPP5P; OCRL2
UniProt ID: (Human) P32019, (Mouse) Q8K337
Entrez Gene ID: (Human) 3633, (Mouse) 16330, (Rat) 362590
Molecular Function: hydrolase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.