Novus Biologicals
Manufacturer Code:NBP189361
Catalog # NBP189361
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:EVVKLIFRESDNDRKVMLQLEKKLFDYFNQEVFRDNNGTALLEFDKELSVFKDRLYELDISFPPSYPYSEDARQGEQYMNTRCPAWCDRILMSPSAKELVLRSESEEKVVTYDHIGPNVCMGDHKPVFL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 43 kDa inositol polyphosphate 5-phophatase; 43 kDa inositol polyphosphate 5-phophatase CTCL tumor antigen HD-CL-025PTASE DKFZp434A1721 EC 3.1.3.56 inositol polyphosphate-5-phosphatase 40kD inositol polyphosphate-5-phosphatase 40kDa inositol trisphosphate-5-phosphatase 40kD InsP3 5-phosphatase MGC1169475PTase MGC116949 type I inositol-145-trisphosphate 5-phophatase type I inositol-145-trisphosphate 5-phosphatase; 43 kDa inositol polyphosphate 5-phosphatase; 5PTase; CTCL tumor antigen HD-CL-02; Inositol polyphosphate-5-phosphatase A; inositol polyphosphate-5-phosphatase, 40kD; inositol polyphosphate-5-phosphatase, 40kDa; inositol trisphosphate-5-phosphatase, 40kD; InsP3 5-phosphatase; Type I inositol 1,4,5-trisphosphate 5-phosphatase; type I inositol-1,4,5-trisphosphate 5-phophatase; type I inositol-1,4,5-trisphosphate 5-phosphatase
Gene Aliases: 5PTASE; INPP5A
UniProt ID: (Human) Q14642
Entrez Gene ID: (Human) 3632
Molecular Function: hydrolase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.