Novus Biologicals
Manufacturer Code:NBP152933
Catalog # NBP152933
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to IMPDH1(IMP (inosine monophosphate) dehydrogenase 1) The peptide sequence was selected from the middle region of IMPDH1. Peptide sequence KKGKLPIVNDCDELVAIIARTDLKKNRDYPLASKDSQKQLLCGAAVGTRE. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp781N0678 EC 1.1.1.205 IMP (inosine 5'-monophosphate) dehydrogenase 1 IMP (inosine monophosphate) dehydrogenase 1 IMP dehydrogenase 1 IMPD IMPD 1 IMPD1 IMPDH 1 IMPDH-I inosine-5'-monophosphate dehydrogenase 1 LCA11RP10 retinitis pigmentosa 10 (autosomal dominant) sWSS2608; IMP (inosine 5'-monophosphate) dehydrogenase 1; IMP (inosine monophosphate) dehydrogenase 1; IMP dehydrogenase 1; IMPD 1; IMPDH 1; IMPDH-I; Inosine-5'-monophosphate dehydrogenase 1
Gene Aliases: IMPD; IMPD1; IMPDH-I; IMPDH1; LCA11; RP10; sWSS2608
UniProt ID: (Human) P20839
Entrez Gene ID: (Human) 3614
Molecular Function: dehydrogenase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.