Proteintech
Manufacturer Code:257391AP150UL
Catalog # 50-173-5049
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | IMPAD1 Fusion Protein Ag22629 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3'(2'), 5'-bisphosphate nucleotidase 2; Golgi 3-prime phosphoadenosine 5-prime phosphate 3-prime phosphatase; Golgi-resident adenosine 3',5'-bisphosphate 3'-phosphatase; golgi-resident nucleotide phosphatase; Golgi-resident PAP phosphatase; IMP 3; IMP 3 IMPA3 IMPAD1 IMPase 3 Inositol monophosphatase 3; IMPase 3; inositol monophosphatase 3; Inositol monophosphatase domain-containing protein 1; inositol-1(or 4)-monophosphatase 3; Myo-inositol monophosphatase A3; Phosphoadenosine phosphate 3'-nucleotidase
Gene Aliases: 1110001C20Rik; AA408880; AI451589; AL022796; B230207P20; BPNT2; GPAPP; IMP 3; IMP-3; IMPA3; IMPAD1; Jaws
UniProt ID: (Human) Q9NX62, (Mouse) Q80V26
Entrez Gene ID: (Human) 54928, (Mouse) 242291
Molecular Function: hydrolase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.