Invitrogen
Manufacturer Code:PA536522
Catalog # PIPA536522
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 220-265 of human IL-4R alpha |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD124; IL-4 receptor subunit alpha; IL-4-binding protein; IL4-BP; IL4R nirs variant 1; interleukin-4 receptor alpha chain; Interleukin-4 receptor subunit alpha; Interleukin-4 receptor subunit alpha IL-4 receptor subunit alpha IL-4R subunit alpha IL-4R-alpha IL-4RA CD124 Soluble interleukin-4 receptor subunit alpha Soluble IL-4 receptor subunit alpha Soluble IL-4R-alpha sIL4Ralpha/prot IL-4-binding protein IL4-BP IL4R IL4RA ORF Names: 582J2.1; Soluble IL-4 receptor subunit alpha; Soluble interleukin-4 receptor subunit alpha
Gene Aliases: 582J2.1; CD124; IL-4RA; IL4R; IL4RA
UniProt ID: (Human) P24394
Entrez Gene ID: (Human) 3566
Molecular Function: cytokine receptor receptor type I cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.