Abnova
Manufacturer Code:MAB5807
Catalog # 89-112-471
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 5H4 |
Immunogen | Rat myeloma YB2/0 transfected with truncated mouse IL-2RB cDNA. |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store in the dark at 4°C. Do not freeze.<br />Avoid prolonged exposure to light.<br />Aliquot to avoid repeated freezing and thawing. |
Rat monoclonal antibody raised against Il2rb.
For Research Use Only
Protein Aliases: CD122; High affinity IL-2 receptor subunit beta; IL-15 receptor beta chain; IL-15Rbeta; IL-2 receptor beta chain; IL-2 receptor subunit beta; IL-2/15 receptor-beta; Il-2/15Rbeta; IL-2R subunit beta; IL-2RB; Il-2Rbeta; IL15Rbeta; Interleukin-2 receptor subunit beta; MGC118674; p70; p70-75
Gene Aliases: CD122; IL-15Rbeta; Il-2/15Rbeta; Il-2Rbeta; IL15Rbeta; Il2rb; p70
UniProt ID: (Mouse) P16297
Entrez Gene ID: (Mouse) 16185
Molecular Function: cytokine receptor receptor type I cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.