Invitrogen
Manufacturer Code:MA530804
Catalog # PIMA530804
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 12 |
Immunogen | Recombit Mouse IL-21 Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: CD360; IL-21 receptor; IL-21R; Interleukin-21 Receptor; LR-beta; Lymphocyte receptor beta; Novel cytokine receptor 8; Novel interleukin receptor; NR8
Gene Aliases: Il21r; NILR
UniProt ID: (Mouse) Q9JHX3
Entrez Gene ID: (Mouse) 60504
Molecular Function: cytokine receptor receptor type I cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.