R&D Systems
Manufacturer Code:MAB7473SP
Catalog # MAB7473SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 1140A |
Immunogen | NSO mouse myeloma cell line transfected with mouse IL-7 R alpha/CD127 Glu21-Asp239 Accession # P16872 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: CD127; CD127 antigen; CD127 CD127 antigen CD127ILRA CDW127 IL-7 R alpha IL-7 receptor subunit alpha IL7R IL7R alpha IL-7R subunit alpha IL7RA IL-7RA IL-7R-alpha interleukin 7 receptor interleukin 7 receptor alpha chain interleukin 7 receptor isoform H5-6 interleukin-7 receptor subunit alpha; CDw127; IL-7 receptor subunit alpha; IL-7R subunit alpha; interleukin 7 receptor alpha chain; interleukin 7 receptor isoform H5-6; Interleukin-7 receptor subunit alpha
Gene Aliases: CD127; CDW127; IL-7R-alpha; IL7R; IL7RA; ILRA
UniProt ID: (Human) Q6NWM0
Entrez Gene ID: (Human) 3575
Molecular Function: cytokine receptor receptor type I cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.